CAS RN 754217-65-7
CAS RN 754217-65-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dimethylarsinothioic Acid, TRC
CAS: 754217-65-7 Molecular Formula: C2H7AsOS Molecular Weight (g/mol): 154.06 Synonym: Dimethylarsinothioic O-Acid; IUPAC Name: dimethylarsinothioic S-acid SMILES: O=[As](S)(C)C