missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl sulfoxide, ≥99%, MP Biomedicals™
£36.46 - £183.38
Chemical Identifiers
CAS | 67-68-5 |
---|---|
Molecular Formula | C2H6OS |
Molecular Weight (g/mol) | 78.13 |
MDL Number | MFCD00002089 |
InChI Key | IAZDPXIOMUYVGZ-UHFFFAOYSA-N |
Synonym | dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide |
PubChem CID | 679 |
ChEBI | CHEBI:28262 |
SMILES | CS(C)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11402691
|
MP Biomedicals
0219605525 |
25 mL |
£36.46
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11412691
|
MP Biomedicals
0219605580 |
100 mL |
£82.73
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11422691
|
MP Biomedicals
0219605590 |
0.5 L |
£130.38
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11432691
|
MP Biomedicals
0219605591 |
1 L |
£183.38
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
67-68-5 | |
78.13 | |
IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
679 | |
CS(C)=O |
C2H6OS | |
MFCD00002089 | |
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
CHEBI:28262 |
Specifications
67-68-5 | |
≥99% | |
MFCD00002089 | |
IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
methylsulfinylmethane | |
679 | |
78.1 | |
Dimethyl sulfoxide |
Liquid | |
C2H6OS | |
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
CS(C)=O | |
78.13 | |
CHEBI:28262 | |
≥99% |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Dimethyl Sulfoxide, cell culture reagent
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.