missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl sulfoxide, ≥99%, MP Biomedicals™

Product Code. 11432691 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
0.5 L
1 L
Unit Size:
1 L
100mL
25mL
500mL
Product Code. Quantity unitSize
11432691 1 L 1 L
11402691 25 mL 25mL
11412691 100 mL 100mL
11422691 0.5 L 500mL
4 options
This item is not returnable. View return policy

Product Code. 11432691

Brand: MP Biomedicals 0219605591

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
SMILES CS(C)=O

Specifications

Quantity 1 L
Physical Form Liquid
Assay Percent Range ≥99%
IUPAC Name methanesulfinylmethane
Formula Weight 78.1
Percent Purity ≥99%
Chemical Name or Material Dimethyl sulfoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.