missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl pyridine-2,3-dicarboxylate, 95%

Product Code. 15452369
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15452369 5 g 5g
15462369 25 g 25g
15472369 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15452369 Supplier Thermo Scientific Alfa Aesar Supplier No. H64939.06
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 605-38-9 | C9H9NO4 | 195.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 605-38-9
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.17
MDL Number MFCD00186549
InChI Key YLGIBCYHQZTFQL-UHFFFAOYSA-N
Synonym 2,3-pyridinedicarboxylic acid dimethyl ester, dimethyl quinolinate, dimethyl 2,3-pyridinedicarboxylate, 2,3-bis methoxycarbonyl pyridine, 2,3-pyridinedicarboxylic acid, dimethyl ester, methylquinolinic acid, 2,3-dimethyl ester, 2,3-dimethyl pyridine-2,3-dicarboxylate, pyridine-2,3-dicarboxylic acid dimethyl ester, mqdme, 2,3-dimethyl methylquinolinate
PubChem CID 69058
IUPAC Name dimethyl pyridine-2,3-dicarboxylate
SMILES COC(=O)C1=CC=CN=C1C(=O)OC

Specifications

Melting Point 53°C to 58°C
Quantity 5 g
Beilstein 159846
Formula Weight 195.17
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Dimethyl pyridine-2,3-dicarboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.