accessibility menu, dialog, popup

UserName

CH3OONOOH3C

CAS RN 605-38-9

IUPAC Name: 2,3-dimethyl pyridine-2,3-dicarboxylate
Synonyms: 2,3-dimethyl pyridine-2,3-dicarboxylate
Molecular Weight (g/mol): 195.17
Molecular Formula: C9H9NO4
InChi Key: YLGIBCYHQZTFQL-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CN=C1C(=O)OC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results