missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 1.3400g/mL |
| Boiling Point | 80°C (25.0 mmHg) |
| Flash Point | >110°C |
| Assay Percent Range | 95% min. (Argentometry) |
| Packaging | Glass bottle |
| Linear Formula | (CH3O)2POCl |
| Quantity | 25 g |
| Specific Gravity | 1.34 |
| IUPAC Name | [chloro(methoxy)phosphoryl]oxymethane |
| Formula Weight | 144.49 |
| Show More |
Safety and Handling
Product Identifier
- Dimethyl chlorophosphate
- Danger
- Acute toxicity Category 1
- Acute toxicity Category 2
- Skin corrosion/irritation Category 1 B
- H310-Fatal in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H330-Fatal if inhaled.
- H300-Fatal if swallowed.
- EUH029-Contact with water liberates toxic gas.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P361-Take off immediately all contaminated clothing.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?