accessibility menu, dialog, popup

UserName

CH3OPClOOH3C

CAS RN 813-77-4

IUPAC Name: dimethyl phosphorochloridate
Synonyms: dimethyl phosphorochloridate
Molecular Weight (g/mol): 144.49
Molecular Formula: C2H6ClO3P
InChi Key: NGFFLHMFSINFGB-UHFFFAOYSA-N
SMILES: COP(Cl)(=O)OC
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)