missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl chlorophosphate, 96%

Product Code. 10234914 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10234914 5 g Glass bottle 5g
10753995 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10234914 Supplier Thermo Scientific Acros Supplier No. 426140050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 813-77-4 | C2H6ClO3P | 144.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 813-77-4
Molecular Formula C2H6ClO3P
Molecular Weight (g/mol) 144.49
MDL Number MFCD00117904
InChI Key NGFFLHMFSINFGB-UHFFFAOYSA-N
Synonym dimethyl phosphorochloridate, dimethyl chlorophosphate, phosphorochloridic acid, dimethyl ester, methyl phosphorochloridate, dimethylchlorophosphate, dimethyl chlorophosphonate, chlorodimethoxyphosphine oxide, chlorodimethylphosphate, dimethyl_chlorophosphate, pubchem19321
PubChem CID 101863
SMILES COP(Cl)(=O)OC

Specifications

Density 1.3400g/mL
Boiling Point 80°C (25.0 mmHg)
Flash Point >110°C
Assay Percent Range 95% min. (Argentometry)
Packaging Glass bottle
Linear Formula (CH3O)2POCl
Quantity 5 g
Specific Gravity 1.34
IUPAC Name dimethyl phosphorochloridate
Formula Weight 144.49
Percent Purity 96%
Chemical Name or Material Dimethyl chlorophosphate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dimethyl chlorophosphate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H310-Fatal in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H300-Fatal if swallowed.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361-Take off immediately all contaminated clothing.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.