missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl aminomalonate hydrochloride, 97%

Product Code. 10735413 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10735413 25 g Glass bottle 25g
10210213 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10735413 Supplier Thermo Scientific Acros Supplier No. 374970250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16115-80-3 | C5H10ClNO4 | 183.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16115-80-3
Molecular Formula C5H10ClNO4
Molecular Weight (g/mol) 183.59
MDL Number MFCD00038880
InChI Key QWNDKNJSEWOEDM-UHFFFAOYSA-N
Synonym dimethyl 2-aminomalonate hydrochloride, dimethyl aminomalonate hydrochloride, dimethyl aminomalonate hcl, aminomalonic acid dimethyl ester hydrochloride, 1,3-dimethyl 2-aminopropanedioate hydrochloride, dimethyl 2-aminopropanedioate hydrochloride, acmc-1bp9z, 2-amino-malonic acid dimethyl ester hydrochloride, 2-aminomalonic acid dimethyl ester hydrochloride
PubChem CID 2734846
IUPAC Name dimethyl 2-aminopropanedioate;hydrochloride
SMILES [H+].[Cl-].COC(=O)C(N)C(=O)OC

Specifications

Melting Point 155°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Linear Formula H2NCH(CO2CH3)2·HCl
Quantity 25 g
Formula Weight 183.59
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Dimethyl aminomalonate hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.