accessibility menu, dialog, popup

UserName

HClH3COONH2OOCH3

CAS RN 16115-80-3

IUPAC Name: hydrogen 1,3-dimethyl 2-aminopropanedioate chloride
Synonyms: hydrogen 1,3-dimethyl 2-aminopropanedioate chloride
Molecular Weight (g/mol): 183.59
Molecular Formula: C5H10ClNO4
InChi Key: QWNDKNJSEWOEDM-UHFFFAOYSA-N
SMILES: [H+].[Cl-].COC(=O)C(N)C(=O)OC
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)