missing translation for 'onlineSavingsMsg'
Learn More

Dimethindene, TRC

Product Code. 30386578
Change view
Click to view available options
Quantity:
1 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30386578 100 mg
30371764 1 mg
30371653 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30386578 Supplier Toronto Research Chemicals Supplier No. TRCD460803100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5636-83-9
Molecular Formula C20H24N2
Molecular Weight (g/mol) 292.42
Synonym 1H-Indene-2-ethanamine, N,N-dimethyl-3-[1-(2-pyridinyl)ethyl]-, Pyridine, 2-[1-[2-[2-(dimethylamino)ethyl]inden-3-yl]ethyl]- (6CI,7CI,8CI), N,N-Dimethyl-3-[1-(2-pyridinyl)ethyl]-1H-indene-2-ethanamine, (±)-Dimethindene, 2-(1-[2-(2-Dimethylaminoethyl)inden-3-yl]ethyl)pyridine, 3-[α-(2'-Pyridyl)ethyl]-2-(β-dimethylaminoethyl)indene, Dimethindene, Dimethpyrindene, Dimetindene, Feniallerg, Forhistal
SMILES CC(C1=C(CCN(C)C)Cc2ccccc12)c3ccccn3

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C20H24N2/c1-15(19-10-6-7-12-21-19)20-17(11-13-22(2)3)14-16-8-4-5-9-18(16)20/h4-10,12,15H,11,13-14H2,1-3H3
Formula Weight 292.1939
Chemical Name or Material Dimethindene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.