CAS RN 5636-83-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dimethindene, TRC
CAS: 5636-83-9 Molecular Formula: C20H24N2 Molecular Weight (g/mol): 292.42 Synonym: 1H-Indene-2-ethanamine, N,N-dimethyl-3-[1-(2-pyridinyl)ethyl]-,Pyridine, 2-[1-[2-[2-(dimethylamino)ethyl]inden-3-yl]ethyl]- (6CI,7CI,8CI),N,N-Dimethyl-3-[1-(2-pyridinyl)ethyl]-1H-indene-2-ethanamine,(±)-Dimethindene,2-(1-[2-(2-Dimethylaminoethyl)inden-3-yl]ethyl)pyridine,3-[α-(2'-Pyridyl)ethyl]-2-(β-dimethylaminoethyl)indene,Dimethindene,Dimethpyrindene,Dimetindene,Feniallerg,Forhistal SMILES: CC(C1=C(CCN(C)C)Cc2ccccc12)c3ccccn3