Learn More
Thermo Scientific™ DiluFlux™ Pro Automated Gravimetric Dilutor
Description
Accurately dispense broth medium into suitable containers for food testing using the Thermo Scientific™ DiluFlux™ Pro Automated Gravimetric Dilutor. DiluFlux Pro Dilutors are compact, easy-to-use, and designed to deliver precise quantities of diluents rapidly with the touch of a button, so that you can work more confidently and accurately. When combined with the Thermo Scientific™ Peristaltic Pump and Thermo Scientific™ Homogenizer, your lab bench will be equipped with a flexible automated culture media solution to increase your productivity.
Automate the dilution step of your Thermo Scientific™ Dry-Bags™ Enrichment Media, as well as other formats, for maximum efficiency. The easy-to-use DiluFlux Pro Gravimetric Dilutors allow the simultaneous use of two diluents by having dual dispensing heads that are fed by dual peristaltic pumps mounted into the unit.
- Intuitive: simply press two keys, tare and go, and the DiluFlux Pro Dilutor will automatically dilute your sample to the desired dilution factor
- Accurate: featuring Watson-Marlow™ peristaltic pumps, high-tech flow regulators, and top of the line weighing platforms for trusted reproducibility and repeatability
- Versatile: compatible with many prepared media formats, the easy-to clean, stainless steel DiluFlux Pro Dilutor is an ideal fit for any food testing laboratory
- High-quality: the DiluFlux Pro Dilutors are ISO 7218, ISO 6887-1, FDA BAM-compliant and are made in France under ISO 9001:2008 certification
Not all products are available for sale in all territories. Please inquire.
Specifications
Specifications
| Description | DiluFlux Pro-3 Automated Gravimetric Dilutor w/ simple (non-robotic) dispensing arm |
| For Use With | Dry-Bags |
| Includes | Simple Dispensing Arm |
| Material | Stainless Steel (Case) |
| Model | DiluFlux Pro-3 |
| Dimensions (L x W x H) | 41.5 x 31 x 40 cm |
| Overall Length (Metric) | 41.5 cm |
| Overall Width (Metric) | 31 cm |
| Overall Height (Metric) | 40 cm |
| Shipping Weight (Metric) | 10.0 kg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
