missing translation for 'onlineSavingsMsg'
Learn More

Dilactic Acid, TRC

Product Code. 30383557
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383557 100 mg -
30371420 50 mg -
30383720 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30383557 Supplier Toronto Research Chemicals Supplier No. TRCD459915100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19201-34-4
Molecular Formula C6H10O5
Molecular Weight (g/mol) 162.14
Synonym Dilactylic Acid, 2,2'-Oxydi-propionic Acid, 2,2'-Oxybis[propanoic acid], 2,4-Dimethyl-3-oxapentanedioic Acid
IUPAC Name 2-(1-carboxyethoxy)propanoic acid
SMILES CC(OC(C)C(=O)O)C(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C6H10O5/c1-3(5(7)8)11-4(2)6(9)10/h3-4H,1-2H3,(H,7,8)(H,9,10)
Formula Weight 162.0528
Chemical Name or Material Dilactic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.