CAS RN 19201-34-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dilactic Acid, TRC
CAS: 19201-34-4 Molecular Formula: C6H10O5 Molecular Weight (g/mol): 162.14 Synonym: Dilactylic Acid,2,2'-Oxydi-propionic Acid,2,2'-Oxybis[propanoic acid],2,4-Dimethyl-3-oxapentanedioic Acid IUPAC Name: 2-(1-carboxyethoxy)propanoic acid SMILES: CC(OC(C)C(=O)O)C(=O)O