missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyldichlorosilane, 97%

Product Code. 11935911 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11935911 25 g Glass bottle 25g
11336490 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11935911 Supplier Thermo Scientific Acros Supplier No. 432360250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7751-38-4 | C6H14Cl2Si | 185.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7751-38-4
Molecular Formula C6H14Cl2Si
Molecular Weight (g/mol) 185.16
MDL Number MFCD00054895
InChI Key GSENNYNYEKCQGA-UHFFFAOYSA-N
Synonym diisopropyldichlorosilane, dichlorodiisopropylsilane, silane, dichlorobis 1-methylethyl, dichloro-di propan-2-yl silane, silane, dichlorodiisopropyl, acmc-209pad, dichloro diisopropyl silane, di 2-propyl dichlorosilane, dichlorobis propan-2-yl silane, bis chloranyl-di propan-2-yl silane
PubChem CID 2758384
SMILES CC(C)[Si](Cl)(Cl)C(C)C

Specifications

Color Colorless
Density 1.0260g/mL
Boiling Point 168.0°C to 170.0°C
Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4430 to 1.4450 (20°C, 589nm)
Quantity 25 g
Specific Gravity 1.026
IUPAC Name dichloro-di(propan-2-yl)silane
Formula Weight 185.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Diisopropyldichlorosilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diisopropyldichlorosilane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P232-Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.