accessibility menu, dialog, popup

UserName

CH3H3CSiH3CCH3ClCl

CAS RN 7751-38-4

IUPAC Name: dichlorobis(propan-2-yl)silane
Synonyms: dichlorodiisopropylsilane
Molecular Weight (g/mol): 185.16
Molecular Formula: C6H14Cl2Si
InChi Key: GSENNYNYEKCQGA-UHFFFAOYSA-N
SMILES: CC(C)[Si](Cl)(Cl)C(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results