missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (-)-Diisopropyl-L-malate, 97%

Product Code. 11445067
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11445067 5 g 5g
11455067 25 g 25g
11465067 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11445067 Supplier Alfa Aesar™ Supplier No. B21960.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 83541-68-8
Molecular Formula C10H16O5
Molecular Weight (g/mol) 216.23
MDL Number MFCD00274300
InChI Key DQOODBSVDOAUMP-JTQLQIEISA-L
Synonym diisopropyl s---malate, --diisopropyl-l-malate, 1,4-diisopropyl 2s-2-hydroxybutanedioate, diisopropyl d-malate, s-diisopropyl malate, diisopropyl-s---malate, l-malic acid diisopropyl ester, s-2-hydroxybutanedioic acid diisopropyl ester
PubChem CID 11333435
SMILES CC(C)O[C@@](CC([O-])=O)(C(C)C)C([O-])=O

Specifications

Density 1.055
Boiling Point 236°C to 238°C
Quantity 5 g
IUPAC Name dipropan-2-yl (2S)-2-hydroxybutanedioate
Formula Weight 218.25
Percent Purity 97%
Chemical Name or Material (-)-Diisopropyl-L-malate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.