accessibility menu, dialog, popup

UserName

CAS RN 83541-68-8

IUPAC Name: (2S)-2-(propan-2-yl)-2-(propan-2-yloxy)butanedioate
Synonyms: (2S)-2-isopropoxy-2-isopropylbutanedioate
Molecular Weight (g/mol): 216.23
Molecular Formula: C10H16O5
InChi Key: DQOODBSVDOAUMP-JTQLQIEISA-L
SMILES: CC(C)O[C@@](CC([O-])=O)(C(C)C)C([O-])=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results