missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress DiI

Product Code. 16488748
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488748 10 mg 10mg
16424667 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16488748 Supplier MedChemExpress Supplier No. HYD0083
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress DiI (DiIC18(3)) is a lipophilic carbocyanine fluorescent dye for membrane labeling.

DiI has an absorption maximum at 549 nm and an emission maximum 565 nm. DiI is mildly fluorescent in aqueous suspension, but becomes bright when bound to cell membrane.

Specifications

Content And Storage 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light)
Quantity 10 mg
Shipping Condition Room temperature
Solubility DMSO : 12.5 mg/mL (13.39 mM; ultrasonic and warming and heat to 80°C)
Target HBV
Product Type Dye Reagents
Molecular Weight (g/mol) 933.87 g/mol
CAS 41085-99-8
Synonym DiIC18(3)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.