missing translation for 'onlineSavingsMsg'
Learn More

Dihydrovitamin K1 Diacetate, TRC

Product Code. 30383394
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383394 5 mg 5mg
30380240 100 mg 100mg
30388439 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30383394 Supplier Toronto Research Chemicals Supplier No. TRCD4484985MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 604-87-5
Molecular Formula C35H52O4
Molecular Weight (g/mol) 536.78
Synonym [R-[R*,R*-(E)]]-2-Methyl-3-(3,7,11,15-tetramethyl-2-hexadecenyl)-1,4-naphthalenediol Diacetate, 2-Methyl-3-[(2E,7R,11R)-3,7,11,15-tetramethyl-2-hexadecenyl]-1,4-Naphthalenediol, Diacetate, 2-Methyl-3-phytyl-1,4-Naphthalenediol Diacetate, Dihydrovitamin K1 Diacetate, di-O-Acetyldihydro-vitamin K1
SMILES CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC\C(=C\Cc1c(C)c(OC(=O)C)c2ccccc2c1OC(=O)C)\C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C35H52O4/c1-24(2)14-11-15-25(3)16-12-17-26(4)18-13-19-27(5)22-23-31-28(6)34(38-29(7)36)32-20-9-10-21-33(32)35(31)39-30(8)37/h9-10,20-22,24-26H,11-19,23H2,1-8H3/b27-22+/t25-,26-/m1/s1
Formula Weight 536.3866
Percent Purity >95%
Chemical Name or Material Dihydrovitamin K1 Diacetate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.