CAS RN 604-87-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dihydrovitamin K1 Diacetate, TRC
CAS: 604-87-5 Molecular Formula: C35H52O4 Molecular Weight (g/mol): 536.78 Synonym: [R-[R*,R*-(E)]]-2-Methyl-3-(3,7,11,15-tetramethyl-2-hexadecenyl)-1,4-naphthalenediol Diacetate,2-Methyl-3-[(2E,7R,11R)-3,7,11,15-tetramethyl-2-hexadecenyl]-1,4-Naphthalenediol, Diacetate,2-Methyl-3-phytyl-1,4-Naphthalenediol Diacetate,Dihydrovitamin K1 Diacetate,di-O-Acetyldihydro-vitamin K1 SMILES: CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC\C(=C\Cc1c(C)c(OC(=O)C)c2ccccc2c1OC(=O)C)\C