missing translation for 'onlineSavingsMsg'
Learn More

Dihydrovitamin K1 Diacetate, TRC

Product Code. 30380240
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380240 100 mg 100mg
30383394 5 mg 5mg
30388439 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380240 Supplier Toronto Research Chemicals Supplier No. TRCD448498100MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 604-87-5
Molecular Formula C35H52O4
Molecular Weight (g/mol) 536.78
Synonym [R-[R*,R*-(E)]]-2-Methyl-3-(3,7,11,15-tetramethyl-2-hexadecenyl)-1,4-naphthalenediol Diacetate, 2-Methyl-3-[(2E,7R,11R)-3,7,11,15-tetramethyl-2-hexadecenyl]-1,4-Naphthalenediol, Diacetate, 2-Methyl-3-phytyl-1,4-Naphthalenediol Diacetate, Dihydrovitamin K1 Diacetate, di-O-Acetyldihydro-vitamin K1
SMILES CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC\C(=C\Cc1c(C)c(OC(=O)C)c2ccccc2c1OC(=O)C)\C

Specifikationer

Quantity 100 mg
InChI Formula InChI=1S/C35H52O4/c1-24(2)14-11-15-25(3)16-12-17-26(4)18-13-19-27(5)22-23-31-28(6)34(38-29(7)36)32-20-9-10-21-33(32)35(31)39-30(8)37/h9-10,20-22,24-26H,11-19,23H2,1-8H3/b27-22+/t25-,26-/m1/s1
Formula Weight 536.3866
Percent Purity >95%
Chemical Name or Material Dihydrovitamin K1 Diacetate
Grade HPLC
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.