missing translation for 'onlineSavingsMsg'
Learn More

Dihydrophytol, TRC

Product Code. 30380697
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30380697 250 mg
30377360 50 mg
30371840 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380697 Supplier Toronto Research Chemicals Supplier No. TRCD450458250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 645-72-7
Molecular Formula C20H42O
Molecular Weight (g/mol) 298.55
Synonym 3,7,11,15-Tetramethyl-1-hexadecanol, 3,7,11,15-Tetramethylhexadecanol, Octahydrogeranylgeraniol, Phytanic Alcohol
IUPAC Name 3,7,11,15-tetramethylhexadecan-1-ol
SMILES CC(C)CCCC(C)CCCC(C)CCCC(C)CCO

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C20H42O/c1-17(2)9-6-10-18(3)11-7-12-19(4)13-8-14-20(5)15-16-21/h17-21H,6-16H2,1-5H3
Formula Weight 298.3236
Chemical Name or Material Dihydrophytol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.