CAS RN 645-72-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dihydrophytol, TRC
CAS: 645-72-7 Molecular Formula: C20H42O Molecular Weight (g/mol): 298.55 Synonym: 3,7,11,15-Tetramethyl-1-hexadecanol,3,7,11,15-Tetramethylhexadecanol,Octahydrogeranylgeraniol,Phytanic Alcohol IUPAC Name: 3,7,11,15-tetramethylhexadecan-1-ol SMILES: CC(C)CCCC(C)CCCC(C)CCCC(C)CCO