missing translation for 'onlineSavingsMsg'
Learn More

Dihydrogen hexachloroosmate(IV) hydrate, Premion™, 99.95% (metals basis), Os 37% min

Product Code. 11356568
Change view
Click to view available options
Quantity:
0.5 g
1 g
5 g
Unit Size:
0.5g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11356568 0.5 g 0.5g
11346568 1 g 1g
11336568 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11356568 Supplier Thermo Scientific Alfa Aesar Supplier No. 012178.02
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 27057-71-2 | H2Cl6Os | 404.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27057-71-2
Molecular Formula H2Cl6Os
Molecular Weight (g/mol) 404.93
MDL Number MFCD00011329
InChI Key GCRLVKBHFZOVLQ-UHFFFAOYSA-H
Synonym Hydrogen hexachloroosmate(IV); Chloroosmic acid
IUPAC Name hexachloroosmium
SMILES Cl[Os](Cl)(Cl)(Cl)(Cl)Cl

Specifications

Melting Point (decomposition)
Color Black
Linear Formula H2OsCl6·xH2O
Quantity 0.5 g
UN Number UN3260
Sensitivity Hygroscopic
Formula Weight 404.93 (Anhydrous)
Percent Purity 99.95%
Grade Premion
Concentration or Composition (by Analyte or Components) Os 37% min.
Physical Form Crystalline
Assay (metals basis)
Chemical Name or Material Dihydrogen hexachloroosmate(IV) hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.