accessibility menu, dialog, popup

UserName

OsClClClClClClHH

CAS RN 27057-71-2

IUPAC Name: dihydrogen hexachloroosmiumdiuide
Synonyms: dihydrogen hexachloroosmiumdiuide
Molecular Weight (g/mol): 404.95
Molecular Formula: Cl6H2Os
InChi Key: UTIXXJBEMJZGFB-UHFFFAOYSA-J
SMILES: [H+].[H+].Cl[Os--](Cl)(Cl)(Cl)(Cl)Cl
Grade 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)