missing translation for 'onlineSavingsMsg'
Learn More

Dihydro Lafutidine, TRC

Product Code. 30375055
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30375055 10 mg 10mg
30381843 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30375055 Supplier Toronto Research Chemicals Supplier No. TRCD44898510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118288-14-5
Molecular Formula C22H31N3O4S
Molecular Weight (g/mol) 433.56
Synonym 2-[(2-Furanylmethyl)sulfinyl]-N-[4-[[4-(1-piperidinylmethyl)-2-pyridinyl]oxy]butyl]acetamide
SMILES O=C(CS(=O)Cc1occc1)NCCCCOc2cc(CN3CCCCC3)ccn2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C22H31N3O4S/c26-21(18-30(27)17-20-7-6-14-28-20)23-9-2-5-13-29-22-15-19(8-10-24-22)16-25-11-3-1-4-12-25/h6-8,10,14-15H,1-5,9,11-13,16-18H2,(H,23,26)
Formula Weight 433.2035
Chemical Name or Material Dihydro Lafutidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.