CAS RN 118288-14-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dihydro Lafutidine, TRC
CAS: 118288-14-5 Molecular Formula: C22H31N3O4S Molecular Weight (g/mol): 433.56 Synonym: 2-[(2-Furanylmethyl)sulfinyl]-N-[4-[[4-(1-piperidinylmethyl)-2-pyridinyl]oxy]butyl]acetamide SMILES: O=C(CS(=O)Cc1occc1)NCCCCOc2cc(CN3CCCCC3)ccn2