missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Digoxin Monoclonal Antibody (DG D8-9), Invitrogen™
GREENER_CHOICE

Mouse Monoclonal Antibody

£367.00 - £916.00

Specifications

Antigen Digoxin
Clone DG D8-9
Concentration 0.80 mg/mL
Applications ELISA
Classification Monoclonal
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17880466
View Documents
Invitrogen™
MA5-46140
100 μg
£367.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11513423
GREENER_CHOICE
View Documents
Invitrogen™
MA110809
500 μg
£478.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17820476
View Documents
Invitrogen™
MA5-46171
1 mg
£916.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MA1-10809 detects Digoxin. MA1-10809 has been successfully used in ELISA applications. The MA1-10809 immunogen is Digoxin, conjugated to KLH.

Digoxin is a beta blocker drug used to treat heart problems, such as heart failure, high blood pressure.
Specifications

Specifications

Digoxin
0.80 mg/mL
Monoclonal
Liquid
RUO
PBS with no preservative; pH 7.4
Digoxin, conjugated to KLH.
Primary
-20°C, Avoid Freeze/Thaw Cycles
DG D8-9
ELISA
Unconjugated
Mouse
Chemical
C41H64O14; DGX; Lanoxin
IgG1
Protein G
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.