Learn More
Invitrogen™ Digoxin Monoclonal Antibody (DG D8-9), Invitrogen™
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA5-46140
 View more versions of this product
Product Code. 17880466
  • £367.00 / 100µg
Description
MA1-10809 detects Digoxin. MA1-10809 has been successfully used in ELISA applications. The MA1-10809 immunogen is Digoxin, conjugated to KLH.
Digoxin is a beta blocker drug used to treat heart problems, such as heart failure, high blood pressure.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Digoxin
Monoclonal
0.80 mg/mL
PBS with no preservative; pH 7.4
Mouse
Protein G
RUO
Chemical
Antibody
IgG1
ELISA
DG D8-9
Unconjugated
C41H64O14; DGX; Lanoxin
Digoxin, conjugated to KLH.
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Chemical