missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Digoxigenin Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
£170.00 - £406.00
Specifications
| Antigen | Digoxigenin |
|---|---|
| Concentration | 5.0 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:5000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, In-situ Hybridization |
| Applications | In Situ Hybridization (ISH), Immunohistochemistry (Frozen), Immunohistochemistry, Western Blot, ELISA |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18004646
|
Novus Biologicals
NB100-65495-0.1ML |
0.1 mL |
£170.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18740124
|
Novus Biologicals
NB100-65495 |
£406.00
0.50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Digoxigenin Polyclonal specifically detects Digoxigenin in Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen, In-situ Hybridization.Specifications
| Digoxigenin | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:5000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, In-situ Hybridization | |
| Polyclonal | |
| Purified | |
| RUO | |
| Phosphate buffered saline with 0.09% Sodium Azide | |
| Digoxin-3-CM conjugated to keyhole limpet haemocyanin | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 5.0 mg/mL | |
| In Situ Hybridization (ISH), Immunohistochemistry (Frozen), Immunohistochemistry, Western Blot, ELISA | |
| Unconjugated | |
| Sheep | |
| Non-species specific | |
| DIG | |
| IgG | |
| Protein G purified | |
| Recognizes Digoxigenin, a steroid found in plants that is often attached to oligonucleotides and used as a molecular probe to detect mRNA production. This cross reacts with Digoxin and Digitoxin. Negligible reactivity has been observed with Ouabain. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title