missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Digoxigenin Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NB100-65495-0.1ML
This item is not returnable.
View return policy
Description
Digoxigenin Polyclonal specifically detects Digoxigenin in Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen, In-situ Hybridization.
Specifications
| Digoxigenin | |
| Polyclonal | |
| Unconjugated | |
| Phosphate buffered saline with 0.09% Sodium Azide | |
| Sheep | |
| Protein G purified | |
| RUO | |
| Recognizes Digoxigenin, a steroid found in plants that is often attached to oligonucleotides and used as a molecular probe to detect mRNA production. This cross reacts with Digoxin and Digitoxin. Negligible reactivity has been observed with Ouabain. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| In Situ Hybridization (ISH), Immunohistochemistry (Frozen), Immunohistochemistry, Western Blot, ELISA | |
| 5.0 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:5000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, In-situ Hybridization | |
| DIG | |
| Digoxin-3-CM conjugated to keyhole limpet haemocyanin | |
| 0.1 mL | |
| Primary | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction