missing translation for 'onlineSavingsMsg'
Learn More

Difluoromethylenediphosphonic Acid, TRC

Product Code. 30384190
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384190 25 mg 25mg
30372934 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30384190 Supplier Toronto Research Chemicals Supplier No. TRCD44584525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10596-32-4
Molecular Formula C H4 F2 O6 P2
Molecular Weight (g/mol) 211.98
IUPAC Name [difluoro(phosphono)methyl]phosphonic acid
SMILES OP(=O)(O)C(F)(F)P(=O)(O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/CH4F2O6P2/c2-1(3,10(4,5)6)11(7,8)9/h(H2,4,5,6)(H2,7,8,9)
Formula Weight 211.9451
Chemical Name or Material Difluoromethylenediphosphonic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.