CAS RN 10596-32-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Difluoromethylenediphosphonic Acid, TRC
CAS: 10596-32-4 Molecular Formula: C H4 F2 O6 P2 Molecular Weight (g/mol): 211.98 IUPAC Name: [difluoro(phosphono)methyl]phosphonic acid SMILES: OP(=O)(O)C(F)(F)P(=O)(O)O