missing translation for 'onlineSavingsMsg'
Learn More

Diethylene Glycol Monolaurate (>90%), TRC

Product Code. 30379759
Change view
Click to view available options
Quantity:
1 g
5 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30379759 1 g
30384986 5 g
30382027 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30379759 Supplier Toronto Research Chemicals Supplier No. TRCD4443601G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-20-8
Molecular Formula C16 H32 O4
Molecular Weight (g/mol) 288.42
Synonym Dodecanoic Acid 2-(2-Hydroxyethoxy)ethyl Ester
IUPAC Name 2-(2-hydroxyethoxy)ethyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCCOCCO

Specifications

Quantity 1 g
InChI Formula InChI=1S/C16H32O4/c1-2-3-4-5-6-7-8-9-10-11-16(18)20-15-14-19-13-12-17/h17H,2-15H2,1H3
Formula Weight 288.2301
Chemical Name or Material Diethylene Glycol Monolaurate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.