CAS RN 141-20-8
IUPAC Name:
2-(2-hydroxyethoxy)ethyl dodecanoate
Synonyms:
peg-2 laurate
Molecular Weight (g/mol):
288.43
Molecular Formula:
C16H32O4
InChi Key:
WGIMXKDCVCTHGW-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCC(=O)OCCOCCO
1
–
1
of
1
results
1
Diethylene Glycol Monolaurate (>90%), TRC
CAS: 141-20-8 Molecular Formula: C16 H32 O4 Molecular Weight (g/mol): 288.42 Synonym: Dodecanoic Acid 2-(2-Hydroxyethoxy)ethyl Ester IUPAC Name: 2-(2-hydroxyethoxy)ethyl dodecanoate SMILES: CCCCCCCCCCCC(=O)OCCOCCO