missing translation for 'onlineSavingsMsg'
Learn More

Diethyl dimethylmalonate, 97%, Thermo Scientific Chemicals

Product Code. 10215690 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10215690 10 g Glass bottle 10g
10701745 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10215690 Supplier Thermo Scientific Acros Supplier No. 162660100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1619-62-1
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.22
MDL Number MFCD00009128
InChI Key UELKSYXXNGHTSE-UHFFFAOYSA-N
Synonym diethyl 2,2-dimethylmalonate, diethyl dimethylmalonate, propanedioic acid, dimethyl-, diethyl ester, diethyldimethylmalonate, diethyl dimethyl malonate, 1,3-diethyl 2,2-dimethylpropanedioate, malonic acid, dimethyl-, diethyl ester, 2,2-dimethyl-malonic acid diethyl ester, zlchem 328, pubchem16321
PubChem CID 15382
IUPAC Name diethyl 2,2-dimethylpropanedioate
SMILES CCOC(=O)C(C)(C)C(=O)OCC

Specifications

Color Colorless
Density 0.9900g/mL
Boiling Point 192°C
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C2H5O2CC(CH3)2CO2C2H5
Refractive Index 1.4114 to 1.4134
Quantity 10 g
Beilstein 02, II, 572
Specific Gravity 0.99
Formula Weight 188.22
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Diethyl dimethylmalonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.