accessibility menu, dialog, popup

UserName

CH3OOCH3H3COOH3C

CAS RN 1619-62-1

IUPAC Name: 1,3-diethyl 2,2-dimethylpropanedioate
Synonyms: 1,3-diethyl 2,2-dimethylpropanedioate
Molecular Weight (g/mol): 188.22
Molecular Formula: C9H16O4
InChi Key: UELKSYXXNGHTSE-UHFFFAOYSA-N
SMILES: CCOC(=O)C(C)(C)C(=O)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results