missing translation for 'onlineSavingsMsg'
Learn More

Diethylcarbamyl Chloride 99%, Thermo Scientific Chemicals

Product Code. 10734221 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass Bottle
Unit Size:
100mL
500mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10734221 500 mL Glass Bottle 500mL
10040740 5 mL Glass Bottle 5mL
10520401 100 mL Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10734221 Supplier Thermo Scientific Acros Supplier No. 114205000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 88-10-8 | Molecular Formula: C5H10ClNO | Molecular Weight: 135.591 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-10-8
Molecular Formula C5H10ClNO
Molecular Weight (g/mol) 135.591
MDL Number MFCD00000636
InChI Key OFCCYDUUBNUJIB-UHFFFAOYSA-N
Synonym diethylcarbamoyl chloride, diethylcarbamyl chloride, carbamic chloride, diethyl, diethylcarbamic chloride, carbamoyl chloride, diethyl, diethylchloroformamide, carbamidoyl chloride, diethyl, n,n-diethylchloroformamide, unii-5riv1u6h5w, n,n-diethylcarbamyl chloride
PubChem CID 6916
IUPAC Name N,N-diethylcarbamoyl chloride
SMILES CCN(CC)C(=O)Cl

Especificaciones

Melting Point -32.0°C
Boiling Point 117.0°C to 123.0°C (132.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula (C2H5)2NCOCl
Refractive Index 1.4505 to 1.4525
Beilstein 04, 120
Density 1.0700g/mL
Quantity 500 mL
Flash Point 80°C
Formula Weight 135.59
Percent Purity 99%
Specific Gravity 1.07
Physical Form Liquid
Chemical Name or Material Diethylcarbamyl chloride, 99%
Mostrar más Mostrar menos

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.