accessibility menu, dialog, popup

UserName

CAS RN 88-10-8

IUPAC Name: N,N-diethylcarbamoyl chloride
Synonyms: diethylcarbamoyl-chloride carbamic chloride, diethyl-
Molecular Weight (g/mol): 135.59
Molecular Formula: C5H10ClNO
InChi Key: OFCCYDUUBNUJIB-UHFFFAOYSA-N
SMILES: CCN(CC)C(Cl)=O
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (5)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)