missing translation for 'onlineSavingsMsg'
Learn More

Diethylcarbamyl Chloride 99%, Thermo Scientific Chemicals

Product Code. 10040740 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass Bottle
Unit Size:
100mL
500mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10040740 5 mL Glass Bottle 5mL
10520401 100 mL Glass Bottle 100mL
10734221 500 mL Glass Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10040740 Supplier Thermo Scientific Acros Supplier No. 114200050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-10-8
Molecular Formula C5H10ClNO
Molecular Weight (g/mol) 135.591
MDL Number MFCD00000636
InChI Key OFCCYDUUBNUJIB-UHFFFAOYSA-N
Synonym diethylcarbamoyl chloride, diethylcarbamyl chloride, carbamic chloride, diethyl, diethylcarbamic chloride, carbamoyl chloride, diethyl, diethylchloroformamide, carbamidoyl chloride, diethyl, n,n-diethylchloroformamide, unii-5riv1u6h5w, n,n-diethylcarbamyl chloride
PubChem CID 6916
IUPAC Name N,N-diethylcarbamoyl chloride
SMILES CCN(CC)C(=O)Cl

Specifications

Melting Point -32.0°C
Boiling Point 117.0°C to 123.0°C (132.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula (C2H5)2NCOCl
Refractive Index 1.4505 to 1.4525
Beilstein 04, 120
Density 1.0700g/mL
Quantity 5 mL
Flash Point 80°C
Formula Weight 135.59
Percent Purity 99%
Specific Gravity 1.07
Physical Form Liquid
Chemical Name or Material Diethylcarbamyl chloride, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.