missing translation for 'onlineSavingsMsg'
Learn More

Diethylcarbamyl chloride, 98%

Product Code. 11314499
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11314499 25 g 25g
11437667 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11314499 Supplier Thermo Scientific Alfa Aesar Supplier No. A15701.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 88-10-8 | C5H10ClNO | 135.591 g/mol

It is used in the production of pharma ingredients. It is a reagent in the synthesis of S-Methyl-N,N-diethylthiocarbamate Sulfoxide, which is an oxygenated metabolite of Disulfiram (Antabuse) that is capable of in vitro inactivation of liver mitochondrial aldehyde dehydrogenase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the production of pharma ingredients. It is a reagent in the synthesis of S-Methyl-N,N-diethylthiocarbamate Sulfoxide, which is an oxygenated metabolite of Disulfiram (Antabuse) that is capable of in vitro inactivation of liver mitochondrial aldehyde dehydrogenase.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from bases, water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-10-8
Molecular Formula C5H10ClNO
Molecular Weight (g/mol) 135.591
MDL Number MFCD00000636
InChI Key OFCCYDUUBNUJIB-UHFFFAOYSA-N
Synonym diethylcarbamoyl chloride, diethylcarbamyl chloride, carbamic chloride, diethyl, diethylcarbamic chloride, carbamoyl chloride, diethyl, diethylchloroformamide, carbamidoyl chloride, diethyl, n,n-diethylchloroformamide, unii-5riv1u6h5w, n,n-diethylcarbamyl chloride
PubChem CID 6916
IUPAC Name N,N-diethylcarbamoyl chloride
SMILES CCN(CC)C(=O)Cl

Specifications

Melting Point -32°C
Boiling Point 186°C
Linear Formula (CH3CH2)2NCOCl
Refractive Index 1.451
UN Number UN3265
Beilstein 506687
Solubility Information Reacts with water.
Density 1.07
Quantity 25 g
Flash Point 76°C (168°F)
Formula Weight 135.59
Sensitivity Moisture sensitive
Percent Purity 98%
Chemical Name or Material Diethylcarbamyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.