missing translation for 'onlineSavingsMsg'
Learn More

Diethylcarbamoyl Chloride, TRC

Product Code. 30379289
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30379289 5 g
30372654 25 g
30374582 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30379289 Supplier Toronto Research Chemicals Supplier No. TRCD4439155G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-10-8
Molecular Formula C5H10ClNO
Molecular Weight (g/mol) 135.59
Synonym (Diethylamino)carbonyl Chloride, Diethylcarbamoyl Chloride, Diethylcarbamyl Chloride, N,N-Diethylcarbamoyl Chloride, N,N-Diethylchloroformamide, N-(Chlorocarbonyl)diethylamine, NSC 223067
IUPAC Name N,N-diethylcarbamoyl chloride
SMILES CCN(CC)C(=O)Cl

Specifications

Quantity 5 g
InChI Formula InChI=1S/C5H10ClNO/c1-3-7(4-2)5(6)8/h3-4H2,1-2H3
Formula Weight 135.0451
Chemical Name or Material Diethylcarbamoyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.