missing translation for 'onlineSavingsMsg'
Learn More

Diethylammonium diethyldithiocarbamate, 98%

Product Code. 11448683
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11448683

Brand: Thermo Scientific Alfa Aesar A10458.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1518-58-7 | C9H22N2S2

Diethylammonium diethyldithiocarbamate is used in the synthesis of Dithiocarbamate. It serve as hosts for the specific binding of various metal cations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylammonium diethyldithiocarbamate is used in the synthesis of Dithiocarbamate. It serve as hosts for the specific binding of various metal cations.

Solubility
Soluble in Chloroform, Methanol.

Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Incompatible with strong acids, Strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1518-58-7
Molecular Formula C9H22N2S2
MDL Number MFCD00013146

Specifications

Melting Point 81°C to 84°C
Linear Formula (CH3CH2)2NH2+ (CH3CH2)2NCS2-
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Soluble in Chloroform,Methanol.
Formula Weight 222.42
Percent Purity 98%
Chemical Name or Material Diethylammonium diethyldithiocarbamate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.