CAS RN 1518-58-7
IUPAC Name:
diethyl[sulfanyl(carbonothioyl)]amine; diethylamine
Synonyms:
diethylamine; diethylcarbamodithioic acid
Molecular Weight (g/mol):
222.41
Molecular Formula:
C9H22N2S2
InChi Key:
NBGTWXBPCIHUQD-UHFFFAOYSA-N
SMILES:
CCNCC.CCN(CC)C(S)=S
1
–
4
of
4
results
1
Diethylammonium diethyldithiocarbamate, 98%
CAS: 1518-58-7 Molecular Formula: C9H22N2S2 MDL Number: MFCD00013146
2
Diethylammonium N,N-Diethyldithiocarbamate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Diethylammonium N,N-Diethyldithiocarbamate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.