missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethylammonium diethyldithiocarbamate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Diethylammonium diethyldithiocarbamate is used in the synthesis of Dithiocarbamate. It serve as hosts for the specific binding of various metal cations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diethylammonium diethyldithiocarbamate is used in the synthesis of Dithiocarbamate. It serve as hosts for the specific binding of various metal cations.
Solubility
Soluble in Chloroform, Methanol.
Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Incompatible with strong acids, Strong bases.
Diethylammonium diethyldithiocarbamate is used in the synthesis of Dithiocarbamate. It serve as hosts for the specific binding of various metal cations.
Solubility
Soluble in Chloroform, Methanol.
Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Incompatible with strong acids, Strong bases.
Specifications
Melting Point | 81°C to 84°C |
Linear Formula | (CH3CH2)2NH2+ (CH3CH2)2NCS2- |
Quantity | 100 g |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in Chloroform,Methanol. |
Formula Weight | 222.42 |
Percent Purity | 98% |
Chemical Name or Material | Diethylammonium diethyldithiocarbamate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'