missing translation for 'onlineSavingsMsg'
Learn More

Diethylammonium diethyldithiocarbamate, 98%

Product Code. 11458683
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11458683 25 g 25g
11448683 5 g 5g
11468683 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11458683 Supplier Thermo Scientific Alfa Aesar Supplier No. A10458.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1518-58-7 | C9H22N2S2

Diethylammonium diethyldithiocarbamate is used in the synthesis of Dithiocarbamate. It serve as hosts for the specific binding of various metal cations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylammonium diethyldithiocarbamate is used in the synthesis of Dithiocarbamate. It serve as hosts for the specific binding of various metal cations.

Solubility
Soluble in Chloroform, Methanol.

Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Incompatible with strong acids, Strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1518-58-7
Molecular Formula C9H22N2S2
MDL Number MFCD00013146

Specifications

Melting Point 81°C to 84°C
Linear Formula (CH3CH2)2NH2+ (CH3CH2)2NCS2-
Quantity 25 g
Sensitivity Hygroscopic
Solubility Information Soluble in Chloroform,Methanol.
Formula Weight 222.42
Percent Purity 98%
Chemical Name or Material Diethylammonium diethyldithiocarbamate
compliance-icons
Product Identifier
  • Diethylammonium diethyldithiocarbamate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: N-ethylethanamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.