Learn More
Diethyl ether, Spectrophotometric Grade, 99+%, inhibitor free, Thermo Scientific™
Solvent For U.V. Spectrophotometric and General Use
£0.01
Chemical Identifiers
| CAS | 60-29-7 |
|---|---|
| Molecular Formula | C4H10O |
| Molecular Weight (g/mol) | 74.12 |
| MDL Number | MFCD00011646 |
| InChI Key | RTZKZFJDLAIYFH-UHFFFAOYSA-N |
| Synonym | diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether |
| PubChem CID | 3283 |
| ChEBI | CHEBI:35702 |
| IUPAC Name | ethoxyethane |
| SMILES | CCOCC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
11375307
|
Thermo Scientific Alfa Aesar
040976.K2 |
1L |
£0.01
1 L |
Estimated Shipment: 30-09-2026
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C4H10O | |
| MFCD00011646 | |
| diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
| CHEBI:35702 | |
| CCOCC |
Specifications
| 60-29-7 | |
| 0.713 | |
| 170°C (338°F) | |
| 1.3525 | |
| C4H10O | |
| MFCD00011646 | |
| diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
| RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
| ethoxyethane | |
| 3283 | |
| 74.12 | |
| Diethyl ether |
| Colorless liquid | |
| -45°C (-49°F) | |
| Sweet | |
| >99% | |
| 586.3 hPa (440mm Hg) at 20°C | |
| 1155 | |
| Immiscible with water,miscible with alcohol. | |
| CCOCC | |
| 74.12 | |
| CHEBI:35702 | |
| Spectrophotometric |
Safety and Handling
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P314-P330-P370+P378q-P501c
H224-H302-H305-H335-H336-H373
missing translation for 'dotInformation' : DOT Class: 3, Packaging Group: I
missing translation for 'einecsNumber' : 200-467-2
missing translation for 'rtecsNumber' : KI5775000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.