missing translation for 'onlineSavingsMsg'
Learn More

Diethyl ether, Spectrophotometric Grade, 99+%, inhibitor free

Product Code. 11375307
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11375307 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11375307 Supplier Thermo Scientific Alfa Aesar Supplier No. 040976.K2
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Solvent For U.V. Spectrophotometric and General Use | CAS: 60-29-7 | C4H10O | 74.12 g/mol

Solvent for U.V. spectrophotometric and it is also important as solvent in production of cellulose plastics such as cellulose acetate. It is also common solvent for Grignard reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Melting Point °C to 116°C
Color Colorless
Appearance Colorless liquid
Density 0.713
Quantity 1 L
Flash Point -45°C (-49°F)
Ignition Point 170°C (338°F)
Odor Sweet
Physical Form Liquid
Refractive Index 1.3525
Assay Percent Range >99%
Vapor Pressure 586.3 hPa (440mm Hg) at 20°C
UN Number 1155
Boiling Point 34.6°C
Solubility Information Immiscible with water,miscible with alcohol.
Formula Weight 74.12
Grade Spectrophotometry
Chemical Name or Material Diethyl ether
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.